Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- APOBEC3F
- Entrez Gene ID
-
723812
- Gene Name
- apolipoprotein B mRNA editing enzyme catalytic subunit 3F
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 12522703 - 12535854 on Build Mmul_10
- Gene Symbol
- APOBEC3D
- Entrez Gene ID
-
705996
- Gene Name
- apolipoprotein B mRNA editing enzyme catalytic subunit 3D
- Gene Aliases
-
Apobec3DE
- Chromosome Location
-
Chr.10: 12544160 - 12555513 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JF714487.1 |
- |
- |
1118 |
86 |
JF714488.1 |
- |
- |
1055 |
86 |
View Full Product Details