Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACOT6
Entrez Gene ID
697174
Gene Name
acyl-CoA thioesterase 6
Gene Aliases
EGK_18360
Chromosome Location
Chr.7: 135713715 - 135721140 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171952.1 NP_001165423.1 - 456 103
XM_028850216.1 XP_028706049.1 - 1195 103

View Full Product Details