The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199403.1 | NP_955435.1 | 23 - 24 | 3331 | 97 |
XM_006250164.2 | XP_006250226.1 | 23 - 24 | 3789 | 97 | |
XM_006250166.2 | XP_006250228.1 | 22 - 23 | 3768 | 97 | |
XM_008769657.1 | XP_008767879.1 | 23 - 24 | 3801 | 97 | |
XM_008769658.1 | XP_008767880.1 | 22 - 23 | 3780 | 97 | |
GenBank mRNA | AJ421447.1 | - | 23 - 24 | 3331 | 97 |