Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map1a
Entrez Gene ID
25152
Gene Name
microtubule-associated protein 1A
Gene Aliases
Mtap1a
Chromosome Location
Chr.3: 113251864 - 113272193 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234826.2 XP_006234888.1 1 - 2 343 106
XM_006234827.2 XP_006234889.1 1 - 2 343 106

View Full Product Details