The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017065.1 | NP_058761.1 | 7 - 8 | 1153 | 73 |
GenBank mRNA | AY742860.1 | - | - | 1483 | 73 |
X15468.1 | - | 7 - 8 | 1153 | 73 |