Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mthfsd
Entrez Gene ID
687539
Gene Name
methenyltetrahydrofolate synthetase domain containing
Gene Aliases
-
Chromosome Location
Chr.19: 53025526 - 53034828 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772653.1 XP_008770875.1 3 - 4 428 66

View Full Product Details