The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012201.1 | NP_001012201.1 | 1 - 2 | 293 | 150 |
XM_006242944.2 | XP_006243006.1 | 1 - 2 | 292 | 150 | |
XM_006242946.2 | XP_006243008.1 | 1 - 2 | 292 | 150 | |
XM_006242947.2 | XP_006243009.1 | 1 - 2 | 292 | 150 | |
XM_006242948.2 | XP_006243010.1 | 1 - 2 | 292 | 150 | |
GenBank mRNA | AB114443.1 | - | 1 - 2 | 182 | 150 |
BC078966.1 | - | 1 - 2 | 293 | 150 |