Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc51
Entrez Gene ID
316008
Gene Name
coiled-coil domain containing 51
Gene Aliases
RGD1311466
Chromosome Location
Chr.8: 117812090 - 117830848 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014098.1 NP_001014120.1 3 - 4 525 72
XM_006243788.2 XP_006243850.1 3 - 4 729 72
XM_006243789.2 XP_006243851.1 3 - 4 1303 72
XM_006243790.2 XP_006243852.1 3 - 4 803 72
XM_006243791.2 XP_006243853.1 3 - 4 1107 72
XM_006243792.2 XP_006243854.1 3 - 4 685 72
GenBank mRNA BC087581.1 - 3 - 4 525 72

View Full Product Details