The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047092.1 | NP_001040557.1 | 5 - 6 | 952 | 89 |
XM_006248930.2 | XP_006248992.1 | 8 - 9 | 1488 | 89 | |
XM_006248931.2 | XP_006248993.1 | 7 - 8 | 1375 | 89 | |
XM_006248932.2 | XP_006248994.1 | 6 - 7 | 1785 | 89 | |
XM_006248933.2 | XP_006248995.1 | 6 - 7 | 1702 | 89 | |
XM_008768970.1 | XP_008767192.1 | 6 - 7 | 1490 | 89 | |
GenBank mRNA | AJ879913.1 | - | 5 - 6 | 952 | 89 |
BC160815.1 | - | 5 - 6 | 971 | 89 |