Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdr2l
Entrez Gene ID
360656
Gene Name
cerebellar degeneration-related protein 2-like
Gene Aliases
RGD1306881
Chromosome Location
Chr.10: 103934357 - 103948318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247771.2 XP_006247833.1 2 - 3 649 98
XM_006247772.1 XP_006247834.1 2 - 3 548 98
XM_008768410.1 XP_008766632.1 2 - 3 368 98
XM_340933.7 XP_340934.5 2 - 3 650 98

View Full Product Details