Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Suco
Entrez Gene ID
360863
Gene Name
SUN domain containing ossification factor
Gene Aliases
Dd25
Chromosome Location
Chr.13: 79736485 - 79801574 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199403.1 NP_955435.1 14 - 15 1430 129
XM_006250164.2 XP_006250226.1 14 - 15 1885 129
XM_006250166.2 XP_006250228.1 13 - 14 1864 129
XM_008769657.1 XP_008767879.1 14 - 15 1897 129
XM_008769658.1 XP_008767880.1 13 - 14 1876 129
XM_008769659.1 XP_008767881.1 14 - 15 1897 129
GenBank mRNA AJ421447.1 - 14 - 15 1430 129

View Full Product Details