Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cct8
Entrez Gene ID
288305
Gene Name
chaperonin containing Tcp1, subunit 8 (theta)
Gene Aliases
-
Chromosome Location
Chr.11: 27130172 - 27141881 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105897.2 NP_001099367.2 8 - 9 1068 104
GenBank mRNA CO400743.1 - 2 - 3 261 104
CV076369.1 - 5 - 6 646 104
FQ225223.1 - 8 - 9 1041 104
FQ226011.1 - 8 - 9 1041 104

View Full Product Details