Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nobox
Entrez Gene ID
502759
Gene Name
NOBOX oogenesis homeobox
Gene Aliases
RGD1563929
Chromosome Location
Chr.4: 72749478 - 72757066 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192013.1 NP_001178942.1 6 - 7 733 102
XM_008762912.1 XP_008761134.1 6 - 7 928 102

View Full Product Details