Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Golga5
Entrez Gene ID
299258
Gene Name
golgin A5
Gene Aliases
Ret-II
Chromosome Location
Chr.6: 126350569 - 126378735 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033065.1 NP_001028237.1 5 - 6 1303 87
XM_006240461.2 XP_006240523.1 6 - 7 1558 87
GenBank mRNA AY144587.1 - 5 - 6 1303 87

View Full Product Details