Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cst9l
Entrez Gene ID
362231
Gene Name
cystatin 9-like
Gene Aliases
Cst9
Chromosome Location
Chr.3: 143172691 - 143175504 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108597.1 NP_001102067.1 2 - 3 401 66

View Full Product Details