Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajb11
Entrez Gene ID
360734
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 11
Gene Aliases
LRRGT00084
Chromosome Location
Chr.11: 81741342 - 81757806 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015021.1 NP_001015021.1 8 - 9 1037 77
GenBank mRNA AY387070.1 - 8 - 9 857 77
BC093384.1 - 8 - 9 1037 77

View Full Product Details