Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nip30
Entrez Gene ID
307652
Gene Name
NEFA-interacting nuclear protein NIP30
Gene Aliases
RGD1307433
Chromosome Location
Chr.19: 10827057 - 10856747 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014014.1 NP_001014036.1 7 - 8 800 59
XM_006255110.2 XP_006255172.1 7 - 8 1110 59
XM_006255111.2 XP_006255173.1 7 - 8 877 59
XM_006255114.2 XP_006255176.1 8 - 9 1342 59
GenBank mRNA BC079145.1 - 7 - 8 800 59

View Full Product Details