Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olr68
Entrez Gene ID
293214
Gene Name
olfactory receptor 68
Gene Aliases
ratchr1-160844789-160845757_ORF
Chromosome Location
Chr.1: 168136959 - 168137927 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001000133.1 NP_001000133.1 1 - 1 635 80

View Full Product Details