The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037363.1 | NP_001032440.1 | 1 - 1 | 2057 | 108 |
XM_006236984.2 | XP_006237046.1 | 2 - 2 | 2768 | 108 | |
XM_006236985.2 | XP_006237047.1 | 2 - 2 | 3335 | 108 | |
GenBank mRNA | BC107902.1 | - | 1 - 1 | 2057 | 108 |