Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPS3
Entrez Gene ID
851766
Gene Name
Gene Aliases
GEP2, YDR185C
Chromosome Location
Chr.IV: 831934 - 832473 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180493.1 NP_010471.1 1 - 1 349 83

View Full Product Details