Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAM1
Entrez Gene ID
853010
Gene Name
Gene Aliases
YGR113W
Chromosome Location
Chr.VII: 718893 - 719924 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181242.3 NP_011628.4 1 - 1 953 84

View Full Product Details