Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOF2
Entrez Gene ID
853880
Gene Name
Gene Aliases
YKR010C
Chromosome Location
Chr.XI: 458924 - 461239 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179800.3 NP_012935.3 1 - 1 2247 106

View Full Product Details