Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
AGO3
Entrez Gene ID
100499509
Gene Name
argonaute RISC catalytic component 3
Gene Aliases
EIF2C3
Chromosome Location
Chr.6: 92081763 - 92225811 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194974.1 NP_001181903.1 7 - 8 876 97
XM_021093288.1 XP_020948947.1 - 1142 97
XM_021093289.1 XP_020948948.1 - 1237 97
XM_021093290.1 XP_020948949.1 - 919 97
XM_021093291.1 XP_020948950.1 - 1165 97
GenBank mRNA GQ329716.1 - 7 - 8 876 97

View Full Product Details