Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FPGT
Entrez Gene ID
100513664
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
-
Chromosome Location
Chr.6: 138941832 - 138950340 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204852.1 NP_001191781.1 3 - 4 405 98

View Full Product Details