Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
papss1
Entrez Gene ID
448413
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 1
Gene Aliases
atpsk1, papss, sk1
Chromosome Location
Chr.1: 62389110 - 62431802 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006742.1 NP_001006743.1 2 - 3 275 86
XM_012959851.2 XP_012815305.1 2 - 3 456 86
GenBank mRNA BC075507.1 - 2 - 3 275 86

View Full Product Details