Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- arhgdig
- Entrez Gene ID
-
493242
- Gene Name
- Rho GDP dissociation inhibitor (GDI) gamma
- Gene Aliases
-
rhogdi-3, rhogdi3
- Chromosome Location
-
Chr.9: 9240685 - 9283200 on Build UCB_Xtro_10.0
- Gene Symbol
- act3
- Entrez Gene ID
-
394673
- Gene Name
- actin, striated muscle 3
- Gene Aliases
-
MA, ac100
- Chromosome Location
-
Chr.4: 40809507 - 40814375 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CT025518.2 |
- |
- |
198 |
70 |
View Full Product Details