Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eef1a1
Entrez Gene ID
549446
Gene Name
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases
EF1alpha, ccs-3, ccs3, eef-1, eef1a, eef1a-1, eef1a-s, ef-tu, ef1a, graf-1ef, hngc:16303, leng7, pti1
Chromosome Location
Chr.5: 92444756 - 92447569 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016692.2 NP_001016692.1 3 - 4 667 64
GenBank mRNA BC157768.1 - 3 - 4 662 64
CR760182.2 - 3 - 4 667 64
CR761561.2 - 3 - 4 661 64
CR761905.2 - 3 - 4 619 64

Gene Symbol
LOC101734345
Entrez Gene ID
101734345
Gene Name
elongation factor 1-alpha, somatic form-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016686533: 0 - 2894 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004914511.3 XP_004914568.1 3 - 4 907 64

View Full Product Details