Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gsc
Entrez Gene ID
549458
Gene Name
goosecoid homeobox
Gene Aliases
Xgsc, gsc-a, gsc-b
Chromosome Location
Chr.8: 67383562 - 67386268 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016704.2 NP_001016704.1 1 - 2 467 101
GenBank mRNA BC155019.1 - 2 - 3 415 101
BC170819.1 - 1 - 2 372 101
BC170823.1 - 1 - 2 372 101
CR760138.2 - 1 - 2 467 101

View Full Product Details