Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
coq10b
Entrez Gene ID
549873
Gene Name
coenzyme Q10B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683441: 400603 - 407238 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017119.2 NP_001017119.1 3 - 4 466 81
GenBank mRNA CR761163.2 - 3 - 4 466 81

View Full Product Details