Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cracr2b
Entrez Gene ID
100036622
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
efcab4a
Chromosome Location
Chr.4: 35116399 - 35131049 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097181.1 NP_001090650.1 6 - 7 5823 63
GenBank mRNA BC127278.1 - 6 - 7 5823 63
CR761002.2 - 9 - 10 1142 63

View Full Product Details