Gene Transcripts

Gene Symbol
Immt
Entrez Gene ID
76614
Gene Name
inner membrane protein, mitochondrial
Gene Aliases
1700082C19Rik, D830041H16Rik, HMP, P87, P87/89, P89
Chromosome Location
Chr. 6: 71831061 - 71877381 on Build GRCm38
UniGene ID
Mm.235123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK006977.1 - 4 835
AK164694.1 - 2 714
BC145704.1 - 3 771
BC145706.1 - 3 771

View Full Product Details