Gene Transcripts

Gene Symbol
IRX3
Entrez Gene ID
79191
Gene Name
iroquois homeobox 3
Gene Aliases
IRX-1, IRXB1
Chromosome Location
Chr. 16: 54283300 - 54286772 on Build GRCh38
UniGene ID
Hs.499205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024336.2 NP_077312.2 4 2245
XM_005256139.3 XP_005256196.1 4 2747
GenBank mRNA AY335943.1 - 4 2245
BC034328.1 - 3 926

View Full Product Details