Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 5, 6 | 498 | |
XM_005271736.4 | XP_005271793.2 | 6, 7 | 1528 | ||
XM_011543072.2 | XP_011541374.1 | 6, 7 | 730 | ||
XM_011543073.2 | XP_011541375.2 | 6, 7 | 466 | ||
XM_017018595.1 | XP_016874084.1 | 6, 7 | 447 | ||
XM_017018596.1 | XP_016874085.1 | 4, 5 | 724 | ||
XM_017018597.1 | XP_016874086.1 | 6, 7 | 1537 | ||
GenBank mRNA | AL833062.2 | - | 4, 5 | 300 | |
EF127492.1 | - | 5, 6 | 498 |