Gene Transcripts
- Gene Symbol
- NLRC4
- Entrez Gene ID
- 58484
- Gene Name
- NLR family CARD domain containing 4
- Gene Aliases
- AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF
- Chromosome Location
- Chr. 2: 32224449 - 32265743 on Build GRCh38
- UniGene ID
- Hs.574741
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199138.1 |
NP_001186067.1
|
3, 4 |
515 |
|
NM_001199139.1 |
NP_001186068.1
|
3, 4 |
398 |
|
NM_001302504.1 |
NP_001289433.1
|
3, 4 |
529 |
|
NM_021209.4 |
NP_067032.3
|
3, 4 |
529 |
|
XM_017004619.1 |
XP_016860108.1
|
3, 4 |
515 |
|
GenBank mRNA |
AF376061.1 |
- |
4, 5 |
743 |
|
AK095467.1 |
- |
3, 4 |
515 |
|
AK292673.1 |
- |
3, 4 |
500 |
|
AK314762.1 |
- |
3, 4 |
500 |
|
AY027787.1 |
- |
3, 4 |
530 |
|
AY027788.1 |
- |
3, 4 |
530 |
|
AY027789.1 |
- |
3, 4 |
530 |
|
AY027790.1 |
- |
3 |
530 |
|
AY032589.1 |
- |
2, 3 |
289 |
|
AY035391.1 |
- |
3, 4 |
398 |
|
AY358152.1 |
- |
3, 4 |
477 |
|
BC031555.1 |
- |
3, 4 |
485 |
5179909 |
DN998613.1 |
- |
3, 4 |
535 |
|
View Full Product Details