Gene Transcripts

Gene Symbol
KLF17
Entrez Gene ID
128209
Gene Name
Kruppel like factor 17
Gene Aliases
ZNF393, Zfp393
Chromosome Location
Chr. 1: 44043927 - 44135140 on Build GRCh38
UniGene ID
Hs.567674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173484.3 NP_775755.3 4 2877
GenBank mRNA BC049844.1 - 4 2877

View Full Product Details