Gene Transcripts

Gene Symbol
DUSP15
Entrez Gene ID
128853
Gene Name
dual specificity phosphatase 15
Gene Aliases
C20orf57, VHY
Chromosome Location
Chr. 20: 31845578 - 31870744 on Build GRCh38
UniGene ID
Hs.434108
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012644.2 NP_001012662.1 3, 4 557
NM_001320478.1 NP_001307407.1 3, 4 214
NM_001320479.1 NP_001307408.1 3, 4 214
NM_080611.4 NP_542178.2 3, 4 553
NM_177991.2 NP_817130.1 3, 4 319
XM_017027654.1 XP_016883143.1 3, 4 411
XM_017027655.1 XP_016883144.1 3, 4 202
XM_017027656.1 XP_016883145.1 3, 4 411
XM_017027657.1 XP_016883146.1 3, 4 411
XM_017027658.1 XP_016883147.1 3, 4 351
XM_017027659.1 XP_016883148.1 3, 4 357
XM_017027660.1 XP_016883149.1 3, 4 239
XM_017027661.1 XP_016883150.1 2, 3 348
XM_017027662.1 XP_016883151.1 3, 4 209
GenBank mRNA AK091960.1 - 2, 3 753
AK097430.1 - 3, 4 216
BC056911.1 - 3, 4 370 6195838
BI517878.1 - 3, 4 286
BI560652.1 - 3, 4 557
BI562598.1 - 3, 4 364
BI829823.1 - 3, 4 484
BM554314.1 - 3, 4 179
DN996690.1 - 3, 4 326

View Full Product Details