Gene Transcripts

Gene Symbol
UBE2Q2
Entrez Gene ID
92912
Gene Name
ubiquitin conjugating enzyme E2 Q2
Gene Aliases
Chromosome Location
Chr. 15: 75843286 - 75901047 on Build GRCh38
UniGene ID
Hs.23033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145335.1 NP_001138807.1 6 883
NM_001284382.1 NP_001271311.1 5 897
NM_173469.3 NP_775740.1 6 1002
XM_005254787.1 XP_005254844.1 6 774
XM_017022726.1 XP_016878215.1 6 878
XM_017022727.1 XP_016878216.1 5 697
GenBank mRNA AK000617.1 - 5 876
AK294199.1 - 6 543
AK296234.1 - 6 883
AL832429.1 - 6 827
BC006827.1 - 4 277
BC017708.1 - 6 829 3907760

View Full Product Details