Gene Transcripts
- Gene Symbol
- UBE2G2
- Entrez Gene ID
- 7327
- Gene Name
- ubiquitin conjugating enzyme E2 G2
- Gene Aliases
- UBC7
- Chromosome Location
- Chr. 21: 44768580 - 44801836 on Build GRCh38
- UniGene ID
- Hs.529420
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001202489.1 |
NP_001189418.1
|
3, 4 |
401 |
|
NM_003343.5 |
NP_003334.2
|
5, 6 |
483 |
|
NM_182688.2 |
NP_872630.1
|
6, 7 |
598 |
|
XM_017028469.1 |
XP_016883958.1
|
5, 6 |
442 |
|
GenBank mRNA |
AF032456.1 |
- |
5, 6 |
450 |
|
AK075143.1 |
- |
1, 2 |
624 |
|
AK122700.1 |
- |
1, 2 |
1900 |
|
AK290629.1 |
- |
5, 6 |
483 |
|
AL834129.1 |
- |
1, 2 |
1215 |
|
BC001738.1 |
- |
5, 6 |
445 |
3536853 |
BC008351.1 |
- |
5, 6 |
443 |
3536171 |
BC011569.1 |
- |
5, 6 |
455 |
4548763 |
BE292782.1 |
- |
6, 7 |
537 |
|
BG702453.1 |
- |
5, 6 |
474 |
|
BT006914.1 |
- |
5, 6 |
395 |
|
DA710059.1 |
- |
3, 4 |
392 |
|
View Full Product Details