Gene Transcripts

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
Chromosome Location
Chr. 7: 37848597 - 37900401 on Build GRCh38
UniGene ID
Hs.723454
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016616.4 NP_057700.3 9 864
GenBank mRNA AF202051.2 - 8 593
AF305596.1 - 8 562
AK313599.1 - 8 604
BC036816.1 - 9 864 5745079
GQ472221.1 - 9 864

View Full Product Details