Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256371.1 | NP_001243300.1 | 3 | 413 | |
NM_001256372.1 | NP_001243301.1 | 3 | 358 | ||
NM_024680.3 | NP_078956.2 | 3 | 612 | ||
XM_011520367.1 | XP_011518669.1 | 3 | 614 | ||
GenBank mRNA | AB231783.1 | - | 3 | 300 | |
AK055206.1 | - | 3 | 556 | ||
AK292688.1 | - | 3 | 323 | ||
BC108700.1 | - | 3 | 601 | ||
BC126400.1 | - | 3 | 603 | 8992116 | |
BC126402.1 | - | 3 | 603 | 8992118 | |
BX504614.1 | - | 3 | 358 | ||
DA758300.1 | - | 3 | 413 |