Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005474.2 | NP_001005474.1 | 5 | 531 | |
NM_031419.3 | NP_113607.1 | 4 | 664 | ||
GenBank mRNA | AB037925.1 | - | 4 | 597 | |
AF548362.1 | - | 5 | 528 | ||
AK054787.1 | - | 5 | 400 | ||
AK091782.1 | - | 4 | 664 | ||
BC014587.2 | - | 311 | |||
BC060800.1 | - | 4 | 664 | 30344780 | |
BM050734.1 | - | 5 | 513 | ||
DA541852.1 | - | 5 | 531 | ||
DQ224339.1 | - | 4 | 453 |