Gene Transcripts

Gene Symbol
MFSD7
Entrez Gene ID
84179
Gene Name
major facilitator superfamily domain containing 7
Gene Aliases
LP2561
Chromosome Location
Chr. 4: 676205 - 689468 on Build GRCh38
UniGene ID
Hs.567612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294341.1 NP_001281270.1 4 886
NM_032219.3 NP_115595.2 4 886
XM_005272299.4 XP_005272356.1 4 913
XM_006713918.1 XP_006713981.1 4 540
XM_011513568.1 XP_011511870.1 4 1690
XM_011513571.1 XP_011511873.1 5 710
XM_011513573.2 XP_011511875.1 4 913
XM_017008700.1 XP_016864189.1 4 913
GenBank mRNA AK292010.1 - 4 784
AY358401.1 - 4 588
BC030246.1 - 4 629 5216645

View Full Product Details