Gene Transcripts

Gene Symbol
TSR2
Entrez Gene ID
90121
Gene Name
TSR2, ribosome maturation factor
Gene Aliases
DBA14, DT1P1A10, WGG1
Chromosome Location
Chr. X: 54440420 - 54445297 on Build GRCh38
UniGene ID
Hs.522662
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_058163.1 NP_477511.1 4, 5 451
GenBank mRNA BC007699.1 - 4, 5 451
BC011825.1 - 4, 5 451 3830864
U92980.1 - 3, 4 357

View Full Product Details