Gene Transcripts
- Gene Symbol
- RFTN2
- Entrez Gene ID
- 130132
- Gene Name
- raftlin family member 2
- Gene Aliases
- C2orf11, Raftlin-2
- Chromosome Location
- Chr. 2: 197568224 - 197676050 on Build GRCh38
- UniGene ID
- Hs.591615
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144629.2 |
NP_653230.2
|
2, 3 |
733 |
|
XM_011510595.2 |
XP_011508897.1
|
2, 3 |
923 |
|
XM_011510596.2 |
XP_011508898.1
|
2, 3 |
923 |
|
XM_011510597.2 |
XP_011508899.1
|
2, 3 |
923 |
|
XM_011510598.2 |
XP_011508900.1
|
2, 3 |
923 |
|
XM_017003332.1 |
XP_016858821.1
|
2, 3 |
923 |
|
GenBank mRNA |
AK055136.1 |
- |
2, 3 |
733 |
|
BC093966.1 |
- |
2, 3 |
380 |
7939811 |
BC111069.1 |
- |
2, 3 |
865 |
5764096 |
BC113361.1 |
- |
2, 3 |
380 |
8322413 |
View Full Product Details