Gene Transcripts
- Gene Symbol
- MRPS21
- Entrez Gene ID
- 54460
- Gene Name
- mitochondrial ribosomal protein S21
- Gene Aliases
- MDS016, MRP-S21, RPMS21
- Chromosome Location
- Chr. 1: 150293842 - 150308979 on Build GRCh38
- UniGene ID
- Hs.405880
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018997.3 |
NP_061870.1
|
1, 2 |
607 |
|
NM_031901.5 |
NP_114107.1
|
2, 3 |
171 |
|
GenBank mRNA |
AB049955.1 |
- |
2, 3 |
144 |
|
AF182417.1 |
- |
1, 2 |
116 |
|
AK289467.1 |
- |
2, 3 |
158 |
|
BC004566.2 |
- |
2, 3 |
140 |
3633098 |
BC006554.2 |
- |
1, 2 |
512 |
2819715 |
BI755132.1 |
- |
1, 2 |
564 |
|
HY161691.1 |
- |
2, 3 |
172 |
|
U79258.1 |
- |
1, 2 |
528 |
|
View Full Product Details