Gene Transcripts

Gene Symbol
GUCY2F
Entrez Gene ID
2986
Gene Name
guanylate cyclase 2F, retinal
Gene Aliases
CYGF, GC-F, GUC2DL, GUC2F, RETGC-2, ROS-GC2
Chromosome Location
Chr. X: 109372061 - 109482056 on Build GRCh38
UniGene ID
Hs.123074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001522.2 NP_001513.2 2 503
GenBank mRNA BC144241.1 - 1 298
L37378.1 - 2 526

View Full Product Details