Gene Transcripts

Gene Symbol
NRIP3
Entrez Gene ID
56675
Gene Name
nuclear receptor interacting protein 3
Gene Aliases
C11orf14, NY-SAR-105
Chromosome Location
Chr. 11: 8980576 - 9004049 on Build GRCh38
UniGene ID
Hs.523467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020645.2 NP_065696.1 7 923
GenBank mRNA AK024750.1 - 7 876
AK098694.1 - 7 876
AY211930.1 - 6 620
BC008835.2 - 7 859 4304488

View Full Product Details