Gene Transcripts

Gene Symbol
IFIT5
Entrez Gene ID
24138
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
ISG58, P58, RI58
Chromosome Location
Chr. 10: 89414568 - 89421002 on Build GRCh38
UniGene ID
Hs.252839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012420.2 NP_036552.1 2 819
GenBank mRNA AK312358.1 - 2 652
BC025786.1 - 2 798 5229306
CR457031.1 - 1 588
U34605.1 - 2 705

View Full Product Details