Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014420.2 | NP_055235.1 | 4 | 730 | |
XM_011544488.2 | XP_011542790.1 | 5 | 1305 | ||
XM_017013316.1 | XP_016868805.1 | 5 | 1028 | ||
GenBank mRNA | AB017788.1 | - | 4 | 619 | |
AF177397.1 | - | 4 | 736 | ||
BC107046.2 | - | 4 | 657 | 40003485 | |
BC107047.2 | - | 4 | 657 | 40003487 |