Gene Transcripts

Gene Symbol
RGS9
Entrez Gene ID
8787
Gene Name
regulator of G-protein signaling 9
Gene Aliases
PERRS, RGS9L
Chromosome Location
Chr. 17: 65137338 - 65227703 on Build GRCh38
UniGene ID
Hs.664380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081955.2 NP_001075424.1 18 2062
NM_003835.3 NP_003826.2 18 2071
XM_011525426.2 XP_011523728.1 11 2242
GenBank mRNA AF071476.1 - 18 1935
AF073710.1 - 9 1790
AF493932.1 - 8 1181
AF493933.1 - 18 1868
AK289876.1 - 18 1935
AK290535.1 - 18 1931
AK309061.1 - 18 1952
AY585190.1 - 8 1181
AY585191.1 - 18 1859
BC022504.1 - 18 1929 4801328
BC096843.1 - 8 1184 6519202

View Full Product Details